Anti-CEND1 Rabbit monoclonal antibody unconjugated [clone: EPR3739]
Lieferant: Abcam
ABCAAB113076-100EA 822 EUR
ABCAAB113076-100
Anti-CEND1 Rabbit monoclonal antibody unconjugated [clone: EPR3739]
Antikörper
Type: Primary
Antigen: CEND1 (cell cycle exit and neuronal differentiation 1)
Clonality: monoclonal
Clone: EPR3739
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen