Anti-IgA Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Lieferant: Abcam
ABCAAB112794-1EA 377 EUR
ABCAAB112794-1
Anti-IgA Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Type: Secondary
Antigen: IgA (Cat) (cat IgA)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen