Anti-IgA Sheep Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Lieferant: Abcam
ABCAAB112631-1EA 396 EUR
ABCAAB112631-1
Anti-IgA Sheep Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Type: Secondary
Antigen: Cow IgM
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestellen