Anti-FITC Mouse monoclonal antibody unconjugated [clone: F4/1]

Lieferant: Abcam
ABCAAB112511-250EA 859 EUR
ABCAAB112511-250
Anti-FITC Mouse monoclonal antibody unconjugated [clone: F4/1]
Antikörper
Type: Primary
Antigen: FITC
Clonality: monoclonal
Clone: F4/1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR