Anti-Melanoma Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB112076-100EA 816 EUR
ABCAAB112076-100
Anti-Melanoma Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Melanoma
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human