Anti-EPCAM Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: VU-1D9]

Lieferant: Abcam
ABCAAB112068-50EA 423 EUR
ABCAAB112068-50
Anti-EPCAM Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: VU-1D9]
Antikörper
Type: Primary
Antigen: EPCAM (epithelial cell adhesion molecule)
Clonality: monoclonal
Clone: VU-1D9
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR