Anti-SREBF2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB112046-100EA 882 EUR
ABCAAB112046-100
Anti-SREBF2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SREBF2 (sterol regulatory element binding transcription factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR