Anti-PYCARD Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB111852-50EA 859 EUR
ABCAAB111852-50
Anti-PYCARD Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PYCARD (PYD and CARD domain containing)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR