Anti-TRPC4 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB111841-50EA 819 EUR
ABCAAB111841-50
Anti-TRPC4 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TRPC4 (transient receptor potential cation channel, subfamily C, member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR