Anti-IL17RD Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB111553-50EA 840 EUR
ABCAAB111553-50
Anti-IL17RD Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IL17RD (interleukin 17 receptor D)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR