Anti-Sprouty 1 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB111523-100EA 856 EUR
ABCAAB111523-100
Anti-Sprouty 1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Sprouty 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human