Anti-ITGAX Armenian Hamster monoclonal antibody Peridinin Chlorophyll/Cy5.5® [clone: N418]

Lieferant: Abcam
ABCAAB111469-50EA 379 EUR
ABCAAB111469-50
Anti-ITGAX Armenian Hamster monoclonal antibody Peridinin Chlorophyll/Cy5.5® [clone: N418]
Type: Primary
Antigen: ITGAX (integrin, alpha X (complement component 3 receptor 4 subunit))
Clonality: monoclonal
Clone: N418
Conjugation: Peridinin Chlorophyll/Cy5.5®
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR