Anti-Spectrin beta 5 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB111223-200EA 835 EUR
ABCAAB111223-200
Anti-Spectrin beta 5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Spectrin beta 5
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR