Anti-TNFRSF1A Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB111119-50EA 1230 EUR
ABCAAB111119-50
Anti-TNFRSF1A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TNFRSF1A (tumor necrosis factor receptor superfamily, member 1A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR