Anti-Luteinizing Hormone Mouse monoclonal antibody [clone: 02A01]

Lieferant: Abcam
ABCAAB111000-1EA 739 EUR
ABCAAB111000-1
Anti-Luteinizing Hormone Mouse monoclonal antibody [clone: 02A01]
Type: Primary
Antigen: Luteinizing Hormone
Clonality: monoclonal
Clone: 02A01
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Horse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR