Anti-Histone acetyltransferase MYST3 Goat polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB110945-50EA 840 EUR
ABCAAB110945-50
Anti-Histone acetyltransferase MYST3 Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: Histone acetyltransferase MYST3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR