Anti-CCT6A Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB110905-100EA 796 EUR
ABCAAB110905-100
Anti-CCT6A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CCT6A (chaperonin containing TCP1, subunit 6A (zeta 1))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR