Anti-IL8 Rabbit Monoclonal Antibody [clone: EPR1116]
Lieferant: Abcam
ABCAAB110727-100EA 799 EUR
ABCAAB110727-100
Anti-IL8 Rabbit Monoclonal Antibody [clone: EPR1116]
Antikörper
Type: Primary
Antigen: IL8 (interleukin 8)
Clonality: monoclonal
Clone: EPR1116
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen