Anti-URM1 Rabbit Monoclonal Antibody [clone: EPR4809]

Lieferant: Abcam
ABCAAB110699-100EA 897 EUR
ABCAAB110699-100
Anti-URM1 Rabbit Monoclonal Antibody [clone: EPR4809]
Antikörper
Type: Primary
Antigen: URM1 (ubiquitin related modifier 1)
Clonality: monoclonal
Clone: EPR4809
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR