Anti-HLA B7 Rabbit Monoclonal Antibody [clone: EP2624]
Lieferant: Abcam
ABCAAB110645-100EA 781 EUR
ABCAAB110645-100
Anti-HLA B7 Rabbit Monoclonal Antibody [clone: EP2624]
Antikörper
Type: Primary
Antigen: HLA B7
Clonality: monoclonal
Clone: EP2624
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen