Anti-CD34 Rabbit Monoclonal Antibody [clone: EPR2999]

Lieferant: Abcam
ABCAAB110643-100EA 874 EUR
ABCAAB110643-100
Anti-CD34 Rabbit Monoclonal Antibody [clone: EPR2999]
Antikörper
Type: Primary
Antigen: CD34 (CD34 molecule)
Clonality: monoclonal
Clone: EPR2999
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR