Anti-SYTL4 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB110519-100EA 835 EUR
ABCAAB110519-100
Anti-SYTL4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SYTL4 (synaptotagmin-like 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR