Anti-ZDHHC20 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB110478-100EA 797 EUR
ABCAAB110478-100
Anti-ZDHHC20 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZDHHC20 (zinc finger, DHHC-type containing 20)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR