Anti-DMPO Nitrone Adduct Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: N1664A]

Lieferant: Abcam
ABCAAB110429-100EA 849 EUR
ABCAAB110429-100
Anti-DMPO Nitrone Adduct Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: N1664A]
Antikörper
Type: Primary
Antigen: DMPO Nitrone Adduct
Clonality: monoclonal
Clone: N1664A
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR