Anti-PPIF Mouse Monoclonal Antibody [clone: E11AE12BD4]

Lieferant: Abcam
ABCAAB110324-100EA 981 EUR
ABCAAB110324-100
Anti-PPIF Mouse Monoclonal Antibody [clone: E11AE12BD4]
Antikörper
Type: Primary
Antigen: PPIF (peptidylprolyl isomerase F)
Clonality: monoclonal
Clone: E11AE12BD4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR