Anti-ACO2 Mouse Monoclonal Antibody [clone: 6F12BD9]

Lieferant: Abcam
ABCAAB110321-100EA 912 EUR
ABCAAB110321-100
Anti-ACO2 Mouse Monoclonal Antibody [clone: 6F12BD9]
Antikörper
Type: Primary
Antigen: ACO2 (aconitase 2, mitochondrial)
Clonality: monoclonal
Clone: 6F12BD9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR