Anti-ACADVL Mouse Monoclonal Antibody [clone: 6A9AF2]

Lieferant: Abcam
ABCAAB110285-100EA 864 EUR
ABCAAB110285-100
Anti-ACADVL Mouse Monoclonal Antibody [clone: 6A9AF2]
Antikörper
Type: Primary
Antigen: ACADVL (acyl-CoA dehydrogenase, very long chain)
Clonality: monoclonal
Clone: 6A9AF2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR