Anti-DCXR Mouse monoclonal antibody unconjugated [clone: 4G4AF5]

Lieferant: Abcam
ABCAAB110283-100EA 912 EUR
ABCAAB110283-100
Anti-DCXR Mouse monoclonal antibody unconjugated [clone: 4G4AF5]
Antikörper
Type: Primary
Antigen: DCXR (dicarbonyl/L-xylulose reductase)
Clonality: monoclonal
Clone: 4G4AF5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR