Anti-ATP5H Mouse monoclonal antibody unconjugated [clone: 7F9BG1]

Lieferant: Abcam
ABCAAB110275-100EA 890 EUR
ABCAAB110275-100
Anti-ATP5H Mouse monoclonal antibody unconjugated [clone: 7F9BG1]
Antikörper
Type: Primary
Antigen: ATP5H (ATP synthase, H+ transporting, mitochondrial Fo complex, subunit d)
Clonality: monoclonal
Clone: 7F9BG1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR