Anti-MT-CO1 Mouse Monoclonal Antibody [clone: 11D8B7]

Lieferant: Abcam
ABCAAB110270-100EA 900 EUR
ABCAAB110270-100
Anti-MT-CO1 Mouse Monoclonal Antibody [clone: 11D8B7]
Antikörper
Type: Primary
Antigen: MT-CO1 (mitochondrially encoded cytochrome c oxidase I)
Clonality: monoclonal
Clone: 11D8B7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR