Anti-COX4 + COX4L2 Mouse Monoclonal Antibody [clone: 10G8D12C12]

Lieferant: Abcam
ABCAAB110261-100EA 873 EUR
ABCAAB110261-100
Anti-COX4 + COX4L2 Mouse Monoclonal Antibody [clone: 10G8D12C12]
Antikörper
Type: Primary
Antigen: COX4 + COX4L2
Clonality: monoclonal
Clone: 10G8D12C12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR