Anti-UQCRC1 Mouse Monoclonal Antibody [clone: 16D10AD9AH5]

Lieferant: Abcam
ABCAAB110252-100EA 870 EUR
ABCAAB110252-100
Anti-UQCRC1 Mouse Monoclonal Antibody [clone: 16D10AD9AH5]
Antikörper
Type: Primary
Antigen: UQCRC1 (ubiquinol-cytochrome c reductase core protein I)
Clonality: monoclonal
Clone: 16D10AD9AH5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR