Anti-TAZ Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB110239-100EA 837 EUR
ABCAAB110239-100
Anti-TAZ Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TAZ (tafazzin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR