Anti-EYA4 Goat polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB110148-50EA 840 EUR
ABCAAB110148-50
Anti-EYA4 Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: EYA4 (eyes absent homolog 4 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR