Anti-Complex II Immunocapture Mouse Monoclonal Antibody [clone: 4H12BG12AG2]

Lieferant: Abcam
ABCAAB109865-100EA 856 EUR
ABCAAB109865-100
Anti-Complex II Immunocapture Mouse Monoclonal Antibody [clone: 4H12BG12AG2]
Antikörper
Type: Primary
Antigen: Complex II Immunocapture
Clonality: monoclonal
Clone: 4H12BG12AG2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR