Anti-FMR1 Goat polyclonal antibody unconjugated
Lieferant: Abcam
ABCAAB109741-50EA 859 EUR
ABCAAB109741-50
Anti-FMR1 Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: FMR1 (fragile X mental retardation 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human