Anti-MALT1 Rabbit Monoclonal Antibody [clone: EPR604]
Lieferant: Abcam
ABCAAB109724-100EA 849 EUR
ABCAAB109724-100
Anti-MALT1 Rabbit Monoclonal Antibody [clone: EPR604]
Antikörper
Type: Primary
Antigen: MALT1 (mucosa associated lymphoid tissue lymphoma translocation gene 1)
Clonality: monoclonal
Clone: EPR604
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen