Anti-FNTB Rabbit monoclonal antibody unconjugated [clone: EPR4707]

Lieferant: Abcam
ABCAAB109625-100EA 777 EUR
ABCAAB109625-100
Anti-FNTB Rabbit monoclonal antibody unconjugated [clone: EPR4707]
Antikörper
Type: Primary
Antigen: FNTB (farnesyltransferase, CAAX box, beta)
Clonality: monoclonal
Clone: EPR4707
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR