Anti-TRIM28 Rabbit Monoclonal Antibody [clone: EPR5249]

Lieferant: Abcam
ABCAAB109545-100EA 859 EUR
ABCAAB109545-100
Anti-TRIM28 Rabbit Monoclonal Antibody [clone: EPR5249]
Antikörper
Type: Primary
Antigen: TRIM28 (tripartite motif containing 28)
Clonality: monoclonal
Clone: EPR5249
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR