Anti-VASP Rabbit Monoclonal Antibody [clone: EPR1065]

Lieferant: Abcam
ABCAAB109541-100EA 778 EUR
ABCAAB109541-100
Anti-VASP Rabbit Monoclonal Antibody [clone: EPR1065]
Antikörper
Type: Primary
Antigen: VASP (vasodilator-stimulated phosphoprotein)
Clonality: monoclonal
Clone: EPR1065
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR