Anti-TARDBP Rabbit Monoclonal Antibody [clone: EPR5810]

Lieferant: Abcam
ABCAAB109535-100EA 949 EUR
ABCAAB109535-100
Anti-TARDBP Rabbit Monoclonal Antibody [clone: EPR5810]
Antikörper
Type: Primary
Antigen: TARDBP (TAR DNA binding protein)
Clonality: monoclonal
Clone: EPR5810
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR