Anti-TIMM50 Rabbit Monoclonal Antibody [clone: EPR5784]

Lieferant: Abcam
ABCAAB109527-100EA 859 EUR
ABCAAB109527-100
Anti-TIMM50 Rabbit Monoclonal Antibody [clone: EPR5784]
Antikörper
Type: Primary
Antigen: TIMM50 (translocase of inner mitochondrial membrane 50 homolog (S. cerevisiae))
Clonality: monoclonal
Clone: EPR5784
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR