Anti-TOP2B Rabbit Monoclonal Antibody [clone: EPR5377]
Lieferant: Abcam
ABCAAB109524-100EA 853 EUR
ABCAAB109524-100
Anti-TOP2B Rabbit Monoclonal Antibody [clone: EPR5377]
Antikörper
Type: Primary
Antigen: TOP2B (topoisomerase (DNA) II beta 180kDa)
Clonality: monoclonal
Clone: EPR5377
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen