Anti-NOXA2/p67phox Rabbit Monoclonal Antibody [clone: EPR5065]
Lieferant: Abcam
ABCAAB109523-100EA 866 EUR
ABCAAB109523-100
Anti-NOXA2/p67phox Rabbit Monoclonal Antibody [clone: EPR5065]
Antikörper
Type: Primary
Antigen: NOXA2/p67phox
Clonality: monoclonal
Clone: EPR5065
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen