Anti-H3F3AP6 Rabbit Monoclonal Antibody [clone: EPR362]
Lieferant: Abcam
ABCAAB109520-100EA 1050 EUR
ABCAAB109520-100
Anti-H3F3AP6 Rabbit Monoclonal Antibody [clone: EPR362]
Antikörper
Type: Primary
Antigen: H3F3AP6 (H3 histone, family 3A, pseudogene 6)
Clonality: monoclonal
Clone: EPR362
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen