Anti-IKBKB Rabbit Monoclonal Antibody [clone: EPR5037]

Lieferant: Abcam
ABCAAB109509-100EA 942 EUR
ABCAAB109509-100
Anti-IKBKB Rabbit Monoclonal Antibody [clone: EPR5037]
Antikörper
Type: Primary
Antigen: IKBKB (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta)
Clonality: monoclonal
Clone: EPR5037
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR