Anti-TF Rabbit Monoclonal Antibody [clone: EPR2932]

Lieferant: Abcam
ABCAAB109503-100EA 847 EUR
ABCAAB109503-100
Anti-TF Rabbit Monoclonal Antibody [clone: EPR2932]
Antikörper
Type: Primary
Antigen: TF (transferrin)
Clonality: monoclonal
Clone: EPR2932
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR