Anti-TMEM205 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB109468-200EA 797 EUR
ABCAAB109468-200
Anti-TMEM205 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TMEM205 (transmembrane protein 205)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR