Anti-SHC1 Rabbit Monoclonal Antibody [clone: EP419]

Lieferant: Abcam
ABCAAB109455-100EA 780 EUR
ABCAAB109455-100
Anti-SHC1 Rabbit Monoclonal Antibody [clone: EP419]
Antikörper
Type: Primary
Antigen: SHC1 (SHC (Src homology 2 domain containing) transforming protein 1)
Clonality: monoclonal
Clone: EP419
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR