Anti-Ionotropic Glutamate receptor 4 Rabbit Monoclonal Antibody [clone: EPR2512]

Lieferant: Abcam
ABCAAB109431-100EA 834 EUR
ABCAAB109431-100
Anti-Ionotropic Glutamate receptor 4 Rabbit Monoclonal Antibody [clone: EPR2512]
Antikörper
Type: Primary
Antigen: Ionotropic Glutamate receptor 4
Clonality: monoclonal
Clone: EPR2512
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR